EMAIL THIS PAGE TO A FRIEND
Journal of the American Dental Association (1939)
Acceptance of OMNI-II disinfectant for instruments and equipment. Council on Dental Therapeutics.
PMID 3858354
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
548618
2-Benzyl-4-chlorophenol, 95%
C13H11ClO