EMAIL THIS PAGE TO A FRIEND
Caries research
Determination of salivary Streptococcus mutans level in a stable sucrose-sulphasomidine-containing broth.
PMID 3861253
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46908
Sulfisomidin, VETRANAL, analytical standard
C12H14N4O2S