EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
Lauroyl peroxide.
PMID 3864738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290785
Luperox® LP, Lauroyl peroxide, ≥98%
C24H46O4