EMAIL THIS PAGE TO A FRIEND
Gaoxiong yi xue ke xue za zhi = The Kaohsiung journal of medical sciences
Effects of airborne fluorides on workers at a cryolite production plant.
PMID 3871033
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01713
Cryolite, synthetic, ≥97.0% (from F)
AlF6Na3