EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Possibilities for the therapeutic control of fibrosis.
PMID 3884033
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3134
3-Aminopropionitrile fumarate salt, metabolite
C3H6N2 · 0.5C4H4O4