EMAIL THIS PAGE TO A FRIEND
The Journal of prosthetic dentistry
Effect of surface treatments on the bond strength of etched-metal resin-bonded retainers.
PMID 3884784
Abstract
The effects of surface treatments on the shear bond strength of etched metal to bovine enamel were evaluated. Abraded etched metal without contamination had the highest shear bond, while debonding occurred within the resin interface rather than in the resin to etched metal or resin to enamel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
169919
Isobutyl methacrylate, 97%
C8H14O2