EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Demonstration of a pontine-hippocampal projection containing a ranatensin-like peptide.
PMID 3887223
Abstract
Previous studies have demonstrated a neuronal system containing an immunoreactive ranatensin-like peptide (irRT) in rat brain. The results of the present study describe the ascending projections of a prominent irRT cell group in the pons. A major pontine irRT fiber system was shown to innervate the hippocampus but not the mammillary body.