EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A convenient protein substrate for the determination of protease specificity: reduced and S-3-(trimethylated amino)propylated lysozyme.
PMID 3890609
Abstract
Reduced lysozyme was alkylated with 3-bromopropyltrimethylammonium bromide to give reduced and S-3-(trimethylated amino)propylated lysozyme. It was soluble in a wide range of pH and is suitable as the protein substrate to determine protease specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
347604
(3-Bromopropyl)trimethylammonium bromide, 97%
C6H15Br2N