EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Lens neutral proteinase preparations hydrolyze glutamoyl bonds.
PMID 3894042
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0788
Z-Leu-Leu-Glu β-naphthylamide, ≥98%, powder
C35H44N4O7