EMAIL THIS PAGE TO A FRIEND
Seikagaku. The Journal of Japanese Biochemical Society
[Mechanism of action of cerulenin on fatty acid synthetase and cerulenin resistance in a cerulenin-producing fungus].
PMID 3894530
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2389
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
C12H17NO3