EMAIL THIS PAGE TO A FRIEND
Pharmacology & therapeutics
The inhibition of ribonucleoside diphosphate reductase by hydroxyurea, guanazole and pyrazoloimidazole (IMPY).
PMID 3898152
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D26202
3,5-Diamino-1,2,4-triazole, 98%
C2H5N5