EMAIL THIS PAGE TO A FRIEND
Berliner und Munchener tierarztliche Wochenschrift
[Current findings in the chemotherapy of varroatosis].
PMID 3907622
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45357
Bromopropylate, PESTANAL®, analytical standard
C17H16Br2O3