EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Isolation and properties of beta-glucosidase from Ruminococcus albus.
PMID 3918007
Abstract
An enzyme active against p-nitrophenyl-beta-D-glucoside was purified from logarithmic-phase cells of Ruminococcus albus cultivated in a medium containing ball-milled cellulose. The purification yielded homogeneous enzyme after an approximately 520-fold increase in specific activity and a 9% yield. The enzyme was identified as a beta-glucosidase because it can hydrolyze cellobiose and cellooligosaccharides to glucose from the nonreducing ends.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5759
4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)
C18H25NO13