EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Human GM-CSF: molecular cloning of the complementary DNA and purification of the natural and recombinant proteins.
PMID 3923623
Abstract
Clones of complementary DNA encoding the human lymphokine known as granulocyte-macrophage colony-stimulating factor (GM-CSF) were isolated by means of a mammalian cell (monkey COS cell) expression screening system. One of these clones was used to produce recombinant GM-CSF in mammalian cells. The recombinant hematopoietin was similar to the natural product that was purified to apparent homogeneity from medium conditioned by a human T-cell line. The human T-cell GM-CSF was found to be 60 percent homologous with the GM-CSF recently cloned from murine lung messenger RNA.