EMAIL THIS PAGE TO A FRIEND
Clinical genetics
Glycosphingolipid studies of visceral tissues and brain from type 1 Gaucher disease variants.
PMID 3924448
Abstract
Glucosylceramide and glucosylsphingosine isolated from spleen, liver and brain were quantitated and characterized in two unrelated patients with Gaucher disease, neither of whom had clinical or neuropathologic evidence of neuronal involvement. Visceral glucosylceramide accumulation did not differ in the two patients. Hepatic glucosylsphingosine content was 2-fold greater in a young severely affected 3-year-old American Black patient compared to that in a 56-year-old Ashkenazi Jewish patient. In contrast, significant differences in glycosphingolipid content and composition were observed in the brains of these two cases. Cerebral and cerebellar cortical glucosylceramide accumulated to a greater extent (3-fold) in the severely affected 3-year-old patient compared to that in the older case. The compositions of the acyl and sphingosyl base residues of glucosylceramide in the cerebral and cerebellar cortices from the Ashkenazi Jewish patient were similar to those in normal individuals. In comparison, the gray matter glucosylceramide in the severely affected patient had increased percentages of stearic acid (18:0) and eicosasphingenine (d20:1), suggesting that the accumulated substrate was derived from the brain ganglioside pool. Glucosylsphingosine was found in large amounts only in cerebral and cerebellar cortices from the severely affected patient. The glycolipid content and composition in this patient was similar to that found in the Norrbottnian (Type 3) form of Gaucher disease. The differences in glucosylceramide acyl and sphingosyl base composition in gray matter from the severely affected patient and that in the Ashkenazi Jewish patient suggested that the accumulated substrates were metabolized differently by the residual enzymes in each case.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43659
Glucosylsphingosine, ≥98.0% (TLC)
C24H47NO7