EMAIL THIS PAGE TO A FRIEND
Annual review of biochemistry
ADP-ribosylation.
PMID 3927821
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0996 PARP-1 human, recombinant, expressed in E. coli