EMAIL THIS PAGE TO A FRIEND
La Revue du praticien
[Antiviral drugs].
PMID 3937215
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7000
5-Iodo-2′-deoxycytidine
C9H12IN3O4
278610
Moroxydine hydrochloride, 99%
C6H13N5O · HCl