EMAIL THIS PAGE TO A FRIEND
Nature
Molecular structure of an aspartic proteinase zymogen, porcine pepsinogen, at 1.8 A resolution.
PMID 3941737
Abstract
The only well-understood mechanism of zymogen activation is that of the serine proteinases, in which proteolytic cleavage leads to conformational changes resulting in a functional active site. A different mechanism is now unveiled by the crystal structure of pepsinogen. Salt bridges that stabilize the positioning of the N-terminal proenzyme segment across the active site of pepsin are disrupted at low pH, releasing the amino-terminal segment and thereby exposing the catalytic apparatus and the substrate-binding sites.