EMAIL THIS PAGE TO A FRIEND
Chemotherapy
Response of preadult Necator americanus to some known anthelmintics in hamsters.
PMID 3948582
Abstract
Hamsters infected with laboratory-adapted preadult Necator americanus were dosed with 6 reference anthelmintics. Their efficacy was measured in terms of percentage cure of infected animals as well as percentage worm reduction following treatment. Mebendazole and pyrantel were equally effective in this system. Other anthelmintics, including anti-hookworm compound, bephenium hydroxynaphthoate, were less effective. The comparative results revealed that the N. americanus model is sensitive and reliable for identifying and characterizing new anti-parasite preparations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5650
Bephenium hydroxynaphthoate, analytical standard
C28H29NO4