EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Establishment of the maximum permissible concentrations of norbornene, norbornadiene and dicyclopentadiene in reservoir water].
PMID 3957065
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N32407
Bicyclo[2.2.1]hept-2-ene, 99%
C7H10
B33803
Bicyclo[2.2.1]hepta-2,5-diene, 98%
C7H8