EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Tranexamic acid derivatives with enhanced absorption.
PMID 3959024
Abstract
Derivatives of the antifibrinolytic drug tranexamic acid [trans-4-(aminomethyl)cyclohexanecarboxylic acid] containing one or two tranexamic acid moieties were synthesized. Most of the derivatives have good stability in acidic and neutral solutions but are easily hydrolyzed in plasma. By measuring the amount of tranexamic acid excreted in the urine after an oral dose, relative absorptions of a number of derivatives in the rat were estimated. Most of the derivatives showed greater absorption than tranexamic acid itself. 1-[(Ethoxycarbonyl)oxy]ethyl trans-4-(amino-methyl)cyclohexanecarboxylate hydrochloride was chosen for studies in man.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
23030
1-Chloroethyl ethyl carbonate, ≥98.0% (GC)
C5H9ClO3