EMAIL THIS PAGE TO A FRIEND
Mutation research
Genetic toxicological testing of some dyes by the micronucleus test.
PMID 3960045
Abstract
Three widely used dyes, acridine orange, blue VRS and fast green FCF were administered to male mice in order to study the induction of gross chromosomal anomalies using the micronucleus test. All 3 compounds were shown to be clastogenic.
Related Materials