EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[Antibacterial effect of an ammonium salt of 11-aminoundecanoic acid. 5. Organic ammonium salts].
PMID 396536
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A82605
11-Aminoundecanoic acid, 97%
C11H23NO2