EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Absolute configuration of trans-7,8-dihydroxy-7,8-dihydro-7-methylbenzo[a]pyrene enantiomer and the unusual quasidiequatorial conformation of the diacetate and dimenthoxyacetate derivatives.
PMID 3971469
Abstract
The enantiomers of trans-7,8-dihydroxy-7,8-dihydro-7-methylbenzo[a]pyrene (7-MBaP 7,8-dihydrodiol) and of trans-7,8-dihydroxy-7,8,9,10-tetrahydro-7-methylbenzo[a]pyrene (7-MBaP 7,8-tetrahydrodiol) were directly resolved by high-performance liquid chromatography (HPLC) using a commercially available column packed with an (R)-N-(3,5-dinitrobenzoyl)-phenylglycine derivative of gamma-aminopropylsilanized silica. The absolute configurations of the resolved enantiomers were determined by the exciton chirality method. Circular dichroism (CD) spectral analysis of the quasidiequatorial benzo[a]pyrene 7R,8R-dihydrodiol enantiomer and its diacetate and dimenthoxyacetate derivatives indicated conformational changes were induced upon derivatization. However, the characteristic CD Cotton effects of the quasidiequatorial 7-MBaP 7,8-dihydrodiol and its diacetate and dimenthoxyacetate derivatives were similar indicating that the conformation of 7-MBaP trans-7,8-dihydrodiol was not altered upon derivatization. Proton nuclear magnetic resonance (NMR) spectral analyses confirmed that 7-MBaP 7,8-dihydrodiol, its diacetate and dimenthoxyacetate derivatives all have quasidiequatorial conformations. The results indicate that the methyl substituent of 7-MBaP 7,8-dihydrodiol maintains a quasiaxial position regardless of the size of the acyl derivatives linked to the hydroxyl groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
380903
7-Methylbenzo[a]pyrene, 98%
C21H14