EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Leminorella, a new genus of Enterobacteriaceae: identification of Leminorella grimontii sp. nov. and Leminorella richardii sp. nov. found in clinical specimens.
PMID 3972991
Abstract
Leminorella is proposed as a new genus for the group of Enterobacteriaceae formerly known as Enteric Group 57. Strains of Leminorella gave positive tests for H2S production, acid production from L-arabinose and D-xylose, and tyrosine clearing; they were negative for indole production, Voges-Proskauer, urea hydrolysis, phenylalanine deaminase, motility, gelatin liquefaction, lysine and ornithine decarboxylases, arginine dihydrolase, growth in KCN, and acid production from adonitol, D-arabitol, cellobiose, erythritol, D-galactose, myo-inositol, lactose, maltose, D-mannitol, D-mannose, melibiose, alpha-CH3-glucoside, raffinose, L-rhamnose, salicin, D-sorbitol, sucrose, and trehalose. By DNA hybridization, strains of Leminorella were only 3 to 16% related to other Enterobacteriaceae and were divided into three groups. Leminorella grimontii is proposed as the type species for the genus and strain CDC 1944-81, ATCC 33999, is designated as the type strain. There were four strains of L. grimontii from stool specimens and two from urine specimens. L. richardii is proposed as the name for the second species (type strain, CDC 0978-82, ATCC 33998). All four L. richardii strains were from stool specimens. L. grimontii can be distinguished from L. richardii because it produces gas from glucose (100%) and acid from dulcitol (83%) and is methyl red positive (100%). One strain, CDC 3346-72, was more related to L. grimontii by DNA hybridization than to L. richardii, but the lower relatedness to both of these species indicated that it may be a third species. Biochemically it could not be distinguished from L. grimontii. All Leminorella strains were resistant (no zone of inhibition) to ampicillin, carbenicillin, and cephalothin. Some of the Leminorella strains were sent to us for Salmonella serotyping, and two reacted weakly in Salmonella antisera. The clinical significance of Leminorella is unknown.