EMAIL THIS PAGE TO A FRIEND
Journal of the Royal Society of Medicine
Dangers of Clinitest ingestion.
PMID 3973895
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11945 Benedict’s reagent, for the qualitative determination of sugars