EMAIL THIS PAGE TO A FRIEND
Radiobiologiia
[Modification of the action of ionizing radiation on cells of transplantable tumors by administering xanthobine].
PMID 3975364
Abstract
In experiments on mice it was established that the radiosensitizing effect of xanthobin was manifested by the enhancement of the inhibiting action of ionizing radiation on the growth of adenocarcinoma RL-67, diminution of the "shoulder" of the survival curve of Ehrlich ascites tumor cells, and increase in the average life of tumor-bearing animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
775789
8-Bromocaffeine, 98%
C8H9BrN4O2