EMAIL THIS PAGE TO A FRIEND
Radiobiologiia
[Stimulating effect of aminoethylisothiuronium on the immune response and interferonogenesis in irradiated mice].
PMID 3975380
Abstract
Aminoethylisothiuronium (AET) stimulated the formation of antibodies against sheep erythrocytes, not against E. coli, in X-irradiated (4 Gy) mice. The serum containing AET-induced interferon had the same effect. AET also promoted the rejection of the allogenic skin graft in mice irradiated with the same dose. In addition, AET and cystaphos stimulated the induction of interferon by the Newcastle disease virus in mice exposed to doses of 4, 5 or 6 Gy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8397
Cysteamine S-phosphate sodium salt, analytical standard
C2H7NNaO3PS