EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Identification of 2,4-dichloroanisole and 2,4-dichlorophenol as soil degradation products of ring-labelled [14C]2,4-D.
PMID 3978252
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533661
2,4-Dichloroanisole, 97%
C7H6Cl2O