EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Methodology for the determination of dinitroaniline herbicides in tissue and excreta.
PMID 3981970
Abstract
A method is presented for the analysis of trace amounts of dinitroaniline herbicides in tissue and excreta. The method employs extraction of the tissue or excreta with organic solvent, clean up by liquid/liquid partitioning or silica gel chromatography, and ultimate analysis by gas chromatography using electron capture detection. Recoveries of greater than 80% were noted over the range of concentration 0.1 to 1.0 microgram/g.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36528
Butralin, PESTANAL®, analytical standard
C14H21N3O4
45500
Fluchloralin, PESTANAL®, analytical standard
C12H13ClF3N3O4