EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
High-performance liquid chromatographic assay of 6-methylthioguanine, a major metabolite of 6-thioguanine, in urine.
PMID 3988862
Abstract
A flow-fluorimetric high-performance liquid chromatographic assay for 6-methylthioguanine in urine has been developed. This compound is a major catabolite of 6-thioguanine, an important drug in cancer chemotherapy. The metabolite was extracted from alkaline urine with ethyl acetate which was injected onto a reversed-phase high-performance liquid chromatographic system for separation and detection. The method is simple, rapid and sensitive to below 500 ng ml-1 which is below the levels encountered following a therapeutic dose of 6-thioguanine. Another metabolite was chromatographically separated from 6-methylthioguanine and partially characterised.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9546
2-Amino-6-methylmercaptopurine, ≥95%
C6H7N5S