EMAIL THIS PAGE TO A FRIEND
Revue du rhumatisme et des maladies osteo-articulaires
[Osteomalacia disclosing de Toni-Debré-Fanconi syndrome caused by the ingestion of 3-methylchromone].
PMID 3992155
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
732834
3-Methylchromone
C10H8O2