EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
An essential requirement for ferrous-haemoglobin in the hydrogen peroxide stimulated oxidation of red blood cell membrane lipids.
PMID 3994750
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
671576
N-Phenylhydroxylamine, ≥97.0% (GC)
C6H7NO