EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Purification and characterisation of leukotriene A4 hydrolase from rat neutrophils.
PMID 3995081
Abstract
Leukotriene A4 hydrolase was rapidly and extensively purified from rat neutrophils using anion exchange and gel filtration high-pressure liquid chromatography. The enzyme which converts the allylic epoxide leukotriene A4 to the 5,12-dihydroxyeicosatetraenoic acid leukotriene B4 was localized in the cytosolic fraction and exhibited an optimum activity at pH 7.8 and an apparent Km for leukotriene A4 between 2 X 10(-5) and 3 X 10(-5) M. The purified leukotriene A4 hydrolase was shown to have a molecular weight of 68 000 on sodium dodecylsulfate polyacrylamide gel electrophoresis and of 50 000 by gel filtration. The molecular weight and monomeric native form of this enzyme are unique characteristics which distinguish leukotriene A4 hydrolase from previously purified epoxide hydrolases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5170 Leukotriene A4 Hydrolase human, recombinant, expressed in E. coli