EMAIL THIS PAGE TO A FRIEND
The Journal of the Egyptian Medical Association
Ocular pemphigus.
PMID 400364
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0388
6α-Methyl-11β-hydroxyprogesterone
C22H32O3