EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Synthesis of fluorescent and radiolabeled analogues of phosphatidic acid.
PMID 4006033
Abstract
Procedures for the synthesis of fluorescent and radiolabeled analogues of phosphatidic acid are described. The fluorophore 7-nitrobenzo-2-oxa-1,3-diazole (NBD) was coupled to 6-amino-caproic acid and 12-aminododecanoic acid by reaction of NBD-chloride with the amino acids under mild alkaline conditions at room temperature. 1,2-Dioleoyl-sn-[U-14C]glycerol 3-phosphate was prepared by acylation of sn-[U-14C]glycerol 3-phosphate with oleic acid anhydride using dimethylaminopyridine as the catalyst. This compound was converted to 1-oleoyl-sn-[U-14C]glycerol 3-phosphate by hydrolysis with phospholipase A2. The lysophosphatidic acid was reacylated with NBD-aminocaproyl imidazole or NBD-aminododecanoyl imidazole to form the fluorescent, radiolabeled analogue of phosphatidic acid. Fluorescent, non-radiolabeled analogues of phosphatidic acid were prepared by phospholipase D hydrolysis of fluorescent phosphatidylcholine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159247
12-Aminododecanoic acid, 95%
C12H25NO2