EMAIL THIS PAGE TO A FRIEND
European journal of pediatrics
Severe illness caused by the products of bacterial metabolism in a child with a short gut.
PMID 4018104
Abstract
An 8-year-old boy with a short gut had six episodes of metabolic acidosis and neurological dysfunction over a 1 month period. The neurological features consisted of a depressed conscious state, confusion, aggressive behaviour, slurred speech and ataxia. The organic acid profile of urine demonstrated increased amounts of lactic, 3-hydroxypropionic, 3-hydroxyisobutyric, 2-hydroxyisocaproic, phenyllactic, 4-hydroxyphenylacetic and 4-hydroxyphenyllactic acids. Of the lactic acid 99% was D-lactic acid. The anaerobic gut flora consisted almost entirely of Lactobacilli in unusually large numbers. A course of vancomycin prevented further episodes. A urinary organic acid profile may be diagnostic when a person with a short gut develops metabolic acidosis or an unusual encephalopathy and bacterial metabolites should be considered in other patients with unusual combinations of organic acids in the urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219827
(S)-(−)-2-Hydroxyisocaproic acid, 98%
C6H12O3
219819
2-Hydroxyisocaproic acid, 99%
C6H12O3