EMAIL THIS PAGE TO A FRIEND
In vitro cellular & developmental biology : journal of the Tissue Culture Association
A rapid naphthol yellow S method for measuring the cellular protein content of anchorage cultures.
PMID 4019357
Abstract
A rapid method has been developed for measuring the cellular protein content of mono- and multilayered anchorage cultures. Fixed or air dried cultures are stained for 30 min with 0.2% Naphthol Yellow S (NYS) dissolved in 1% acetic acid. Unbound dye is removed by a series of four 2.5 min washes in 1% acetic acid, and protein-bound dye extracted with 10 mM unbuffered Tris base for spectrophotometric optical density determination at 433 nm. The NYS method exhibited a least-squares correlation coefficient of 0.99997 with the Oyama-Eagle Lowry method.