EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica Suecica
Aminolysis and hydrolysis of formanilide in water solutions. V. Influence of the substituent in para-position.
PMID 4024965
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133329
Formanilide, 99%
C7H7NO