EMAIL THIS PAGE TO A FRIEND
Meditsinskaia radiologiia
[Use of nicotinic acid and midocalm for correcting blood coagulation in patients with radiation edema of the extremities].
PMID 4033386
Abstract
A study was made of the functional activity of platelets, plasmatic coagulation and fibrinolysis in patients with late radiation disorders of the blood and lymph flow during treatment with vasodilative drugs: nicotinic acid and midocalm. These drugs tend to decrease the functional activity of platelets and to raise the blood fibrinolytic activity. Their use is indicated to this group of patients to better their intravascular status.