EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Proteins of the kidney microvillar membrane. Purification and properties of carboxypeptidase P from pig kidneys.
PMID 4038259
Abstract
Carboxypeptidase P has been purified by immunoaffinity chromatography from pig kidneys. A single-step assay with Z-Pro-Met (where Z represents benzyloxycarbonyl) as substrate was used, methionine being determined by using L-amino acid oxidase and horseradish peroxidase. The enzyme constitutes about 1.5% of the kidney microvillar proteins. Triton X-100-solubilized and papain-released forms of the enzyme were isolated. The former had an apparent subunit Mr of 135 000, and the latter form contained two polypeptide chains of Mr 128 000 and 95 000. The undenatured forms were dimeric proteins. In common with other microvillar hydrolases, carboxypeptidase P was a glycoprotein and each subunit contained one Zn atom. MnCl2 (1 mM) in the assay was necessary for maximum activity; in its absence, 0.5 mM-ZnSO4 produced a limited activation, but was inhibitory at higher concentrations. The Km for Z-Pro-Met, in the presence of MnCl2, was 4.1 mM, and the kcat. for freshly prepared enzyme was 1230 min-1. The enzyme lost activity during storage at -20 degrees C. In a limited survey of peptides, hydrolysis was observed only with substrates containing a proline, alanine or glycine residue in the P1 position, and these included angiotensins II and III. The best substrate in this series was Val-Ala-Ala-Phe.