EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Metabolism of 1-nitropyrene and 6-nitrobenzo(a)pyrene by intestinal microflora.
PMID 4040636
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR311
6-Nitrobenzo[a]pyrene, BCR® certified Reference Material
C20H11NO2