EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Effects of a cationic surfactant on the rate of alkaline hydrolysis of p-substituted benzenesulfonyl fluorides.
PMID 4042235
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ALD00003
4-Nitrobenzenesulfonyl fluoride
C6H4FNO4S