EMAIL THIS PAGE TO A FRIEND
Life sciences
Stereospecificity of 2-methylpiperidine binding to a nicotinic up-regulatory site in the rat brain P2 preparation.
PMID 4046738
Abstract
(+/-)-2-Methylpiperidine has a high degree of specificity in enhancing the binding of (-)-[3H]nicotine in the rat brain P2 preparation. (-)- and (+)-2-Methylpiperidine have been resolved. The (+) but not the (-) isomer increased the binding of (-)-[3H]nicotine. The two isomers were equally effective in inhibiting the binding of (-)-[3H]nicotine in high concentrations. These data provide additional support for a stereospecific nicotinic up-regulatory site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M72803
2-Methylpiperidine, 98%
C6H13N
W424401
2-Methylpiperidine, 98%
C6H13N