EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Hydrolysis of acetylsalicylsalicylic acid and salicylsalicylic acid in aqueous solution.
PMID 4053241
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001161 Carbasalate impurity B, European Pharmacopoeia (EP) Reference Standard
C16H12O6