EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Fumarate permeation in rat liver mitochondria: fumarate/malate and fumarate/phosphate translocators.
PMID 4062935
Abstract
Fumarate permeation in isolated rat liver mitochondria was demonstrated by measuring malate and phosphate efflux caused by fumarate added externally to the mitochondrial suspension. The existence of two specific fumarate translocators, fumarate/malate and fumarate/phosphate, is shown here. These carriers are distinguished in the light of different kinetic parameters (Km values are 50 microM and 150 microM, and Vmax values are 17 and 40 nmoles/min X mg mitochondrial protein, respectively) and of differing sensitivity to non-penetrant compounds. Fumarate was found to cause oxaloacetate efflux from mitochondria by means of an indirect process which involves the cooperation of both fumarate/malate and malate/oxaloacetate translocators. Results are discussed in the light of the physiological role played by fumarate translocation in both ureogenesis and aminoacid metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P35200
Phenylsuccinic acid, 98%
C10H10O4