EMAIL THIS PAGE TO A FRIEND
Current problems in dermatology
Single injection adjuvant test.
PMID 4064745
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
316539
1,8-Naphthosultone, 98%
C10H6O3S