EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Experimental study of the gonadotoxic action of 1-phenyl-4-amino-5-chloropyridazole-6].
PMID 4065620
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45385
Chloridazon, PESTANAL®, analytical standard
C10H8ClN3O