EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete
[A bile-chrysoidine-glycerol culture medium and its use in the diagnosis of gram-negative aerobic bacteria, especially Enterobacteriaceae].
PMID 4072296
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27240
Chrysoidine G, for microscopy (Bact., Bot., Vit.)
C12H12N4 · HCl
80028
Chrysoidine G, analytical standard
C12H12N4 · HCl