EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Determination of alpha-amanitin and beta-amanitin in human biological fluids by high-performance liquid chromatography.
PMID 4073484
Abstract
A high-performance liquid chromatographic assay of alpha-amanitin and beta-amanitin in human serum, urine, or stomach washings is described. Sample preparation involves a chemical step with deproteinization and organic solvent treatment, and a selective cleanup and concentration step on reversed-phase prepacked cartridges. Separations are performed on a reversed-phase analytical column under isocratic conditions with uv detection at 280 nm. The method allows the quantitation of alpha- and beta-amanitin separately with a detection limit of 10 ng/ml for both toxins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1304
β-Amanitin from Amanita phalloides, ~90% (HPLC)
C39H53N9O15S